Please login to your online account to display your discounted pricing

PG 01, Tocris™

CFTR mutants potentiator

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material PG 01
Assay Percent Range >98%
Molecular Formula C28H29N3O2
Formula Weight 439.55
Synonym N-Methyl-N-[2-[[4-(1-Methylethyl)phenyl]amino]-2-1H-indole-3-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394350

 
r & d systems
394350
50mg Each for $895.76

394310

 
r & d systems
394310
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 439.55
Quantity 50mg
Molecular Formula C28H29N3O2
CAS 853138-65-5
Synonym N-Methyl-N-[2-[[4-(1-Methylethyl)phenyl]amino]-2-1H-indole-3-acetamide
Recommended Storage Store at -20°C
Assay Percent Range >98%
Chemical Name or Material PG 01
Safety and Handling

Store at -20°C