Please login to your online account to display your discounted pricing

PF 915275, Tocris

Potent and selective 11β-hydroxysteroid dehydrogenase inhibitor

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material PF 915275
Assay Percent Range >98%
Molecular Formula C18H14N4O2S
Formula Weight 350.39
Synonym N-(6-Amino-2-pyridinyl)-4'-cyano-[1, 1'-biphenyl]-4-sulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

329110

 
r & d systems
RDS329110
10mg Each for $195.49

329150

 
r & d systems
RDS329150
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 350.39
Quantity 10mg
Chemical Name or Material PF 915275
Synonym N-(6-Amino-2-pyridinyl)-4'-cyano-[1, 1'-biphenyl]-4-sulfonamide
Assay Percent Range >98%
Molecular Formula C18H14N4O2S
CAS 857290-04-1
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature