Please login to your online account to display your discounted pricing

PF 750, Tocris

Selective FAAH inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material PF 750
Assay Percent Range >98%
Molecular Formula C22H23N3O
Formula Weight 345.44
Synonym N-Phenyl-4-(3-quinolinylmethyl)-1-piperidinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

330750

 
r & d systems
RDS330750
50mg Each for $1,050.42

330710

 
r & d systems
RDS330710
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 345.44
Quantity 50mg
CAS 959151-50-9
Assay Percent Range >98%
Chemical Name or Material PF 750
Molecular Formula C22H23N3O
Synonym N-Phenyl-4-(3-quinolinylmethyl)-1-piperidinecarboxamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature