Please login to your online account to display your discounted pricing

PF 573228, Tocris

Potent and selective FAK inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material PF 573228
Assay Percent Range >98%
Molecular Formula C22H20F3N5O3S
Formula Weight 491.49
Synonym 3, 4-Dihydro-6-[[4-[[[3-(methylsulfonyl)phenyl]methyl]amino]-5-(trifluoromethyl)-2-pyrimidinyl]amino]-2(1H)-quinolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

323910

 
r & d systems
323910
10mg Each for $252.39

323950

 
r & d systems
323950
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 491.49
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C22H20F3N5O3S
Recommended Storage Store at +4°C
Chemical Name or Material PF 573228
Synonym 3, 4-Dihydro-6-[[4-[[[3-(methylsulfonyl)phenyl]methyl]amino]-5-(trifluoromethyl)-2-pyrimidinyl]amino]-2(1H)-quinolinone
CAS 869288-64-2
Safety and Handling

Store at +4°C