Please login to your online account to display your discounted pricing

PF 4981517, Tocris™

Selective CYP3A4 inhibitor

$335.85 - $1,384.48

Chemical Identifiers

Chemical Name or Material PF 4981517
Assay Percent Range >98%
Molecular Formula C26H32N8
Formula Weight 456.59
Synonym (1-Methyl-3-[1-methyl-5-(4-methylphenyl)-1H-pyrazol-4-yl]-4-[(3S)-3-piperidin-1-ylpyrrolidin-1-yl]-1H-pyrazolo[3, 4-d]pyrimidine)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

425210

 
r & d systems
425210
10mg Each for $335.85

425250

 
r & d systems
425250
50mg Each for $1,384.48
Description & Specifications

Specifications

Formula Weight 456.59
Quantity 10mg
CAS 1390637-82-7
Synonym (1-Methyl-3-[1-methyl-5-(4-methylphenyl)-1H-pyrazol-4-yl]-4-[(3S)-3-piperidin-1-ylpyrrolidin-1-yl]-1H-pyrazolo[3, 4-d]pyrimidine)
Recommended Storage Store at +4°C
Molecular Formula C26H32N8
Chemical Name or Material PF 4981517
Assay Percent Range >98%
Safety and Handling

Store at +4°C