Please login to your online account to display your discounted pricing

PF 477736, Tocris™

Selective Chk1 inhibitor

$310.33 - $1,282.40

Chemical Identifiers

Chemical Name or Material PF 477736
Assay Percent Range >99%
Molecular Formula C22H25N7O2
Formula Weight 419.48
Synonym (R)-α-Amino-N-[5,6-dihydro-2-(1-methyl-1H-pyrazol-4-yl)-6-oxo-1H-pyrrolo[4,3,2-ef][2,3]benzodiazepin-8-yl]-cyclohexaneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

427750

 
r & d systems
427750
50mg Each for $1,282.40

427710

 
r & d systems
427710
10mg Each for $310.33
Description & Specifications

Specifications

Formula Weight 419.48
Quantity 50mg
CAS 952021-60-2
Synonym (R)-α-Amino-N-[5,6-dihydro-2-(1-methyl-1H-pyrazol-4-yl)-6-oxo-1H-pyrrolo[4,3,2-ef][2,3]benzodiazepin-8-yl]-cyclohexaneacetamide
Recommended Storage Store at +4°C
Chemical Name or Material PF 477736
Molecular Formula C22H25N7O2
Assay Percent Range >99%
Safety and Handling

Store at +4°C