Please login to your online account to display your discounted pricing

PF 4708671, Tocris™

S6K1 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material PF 4708671
Assay Percent Range >98%
Molecular Formula C19H21F3N6
Formula Weight 390.41
Synonym 2-[[4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl]methyl]-5-(trifluoromethyl)-1H-benzo[d]imidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

403210

 
r & d systems
403210
10mg Each for $252.39

403250

 
r & d systems
403250
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 390.41
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material PF 4708671
Synonym 2-[[4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl]methyl]-5-(trifluoromethyl)-1H-benzo[d]imidazole
Molecular Formula C19H21F3N6
CAS 1255517-76-0
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C