Please login to your online account to display your discounted pricing

PF 3845, Tocris™

Selective FAAH inhibitor

$316.20 - $1,307.92

Chemical Identifiers

Chemical Name or Material PF 3845
Assay Percent Range >99%
Molecular Formula C24H23F3N4O2
Formula Weight 456.46
Synonym N-3-Pyridinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methyl]-1-piperidinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

417510

 
r & d systems
RDS417510
10mg Each for $316.20

417550

 
r & d systems
RDS417550
50mg Each for $1,307.92
Description & Specifications

Specifications

Formula Weight 456.46
Quantity 10mg
Recommended Storage Store at +4°C
Synonym N-3-Pyridinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methyl]-1-piperidinecarboxamide
CAS 1196109-52-0
Assay Percent Range >99%
Chemical Name or Material PF 3845
Molecular Formula C24H23F3N4O2
Safety and Handling

Store at +4°C