Please login to your online account to display your discounted pricing

PF 06465469, Tocris™

Potent inhibitor of interleukin-2 inducible T cell kinase (ITK); Also inhibits BTK

$335.85 - $1,384.48

Chemical Identifiers

Chemical Name or Material PF 06465469
Assay Percent Range >95%
Molecular Formula C30H33N7O2
Formula Weight 523.63
Synonym (R)-3-(1-(1-Acryloylpiperidin-3-yl)-4-amino-1H-pyrazolo[3, 4-d]pyrimidin-3-yl)-N-(3-methyl-4-(1-methylethyl))benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

471010

 
r & d systems
471010
10mg Each for $335.85

471050

 
r & d systems
471050
50mg Each for $1,384.48
Description & Specifications

Specifications

Formula Weight 523.63
Quantity 10mg
CAS 1407966-77-1
Recommended Storage Store at +4°C
Chemical Name or Material PF 06465469
Assay Percent Range >95%
Synonym (R)-3-(1-(1-Acryloylpiperidin-3-yl)-4-amino-1H-pyrazolo[3, 4-d]pyrimidin-3-yl)-N-(3-methyl-4-(1-methylethyl))benzamide
Molecular Formula C30H33N7O2
Safety and Handling

Store at +4°C