missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PF 04885614, Tocris™

Potent and selective NaV1.8 channel blocker; active in vivo

$219.42 - $898.19

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C13H14F3N3O
Formula Weight 285.26
Synonym 1-Methyl-[1-[4-(4-trifluoromethoxy)phenyl]-imidazol-2-yl]-ethanamine
Chemical Name or Material PF 04885614
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
491610 R & D Systems
RDS491610
10mg Each for $219.42
491650 R & D Systems
RDS491650
50mg Each for $898.19
Specifications Description & Specifications

Chemical Identifiers

>98%
285.26
PF 04885614
C13H14F3N3O
1-Methyl-[1-[4-(4-trifluoromethoxy)phenyl]-imidazol-2-yl]-ethanamine

Specifications

10mg
C13H14F3N3O
1-Methyl-[1-[4-(4-trifluoromethoxy)phenyl]-imidazol-2-yl]-ethanamine
>98%
285.26
PF 04885614

Safety and Handling

Recommended Storage : Store at +4°C