Please login to your online account to display your discounted pricing

PF 04691502, Tocris™

Potent and selective dual PI 3-K/mTOR inhibitor

$330.06 - $1,362.21

Chemical Identifiers

Chemical Name or Material PF 04691502
Assay Percent Range >98%
Molecular Formula C22H27N5O4
Formula Weight 425.48
Synonym 2-Amino-8-[trans-4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxy-3-pyridinyl)-4-methyl-pyrido[2, 3-d]pyrimidin-7(8H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

482050

 
r & d systems
RDS482050
50mg Each for $1,362.21

482010

 
r & d systems
RDS482010
10mg Each for $330.06
Description & Specifications

Specifications

Formula Weight 425.48
Quantity 50mg
Molecular Formula C22H27N5O4
Recommended Storage Store at -20°C
Chemical Name or Material PF 04691502
Assay Percent Range >98%
CAS 1013101-36-4
Synonym 2-Amino-8-[trans-4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxy-3-pyridinyl)-4-methyl-pyrido[2, 3-d]pyrimidin-7(8H)-one
Safety and Handling

Store at -20°C