missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PF 04217903 mesylate, Tocris™

Highly selective c-Met inhibitor

$313.93 - $1,298.51

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C19H16N8O·CH3SO3H
Formula Weight 468.49
Synonym 4-[1-(6-Quinolinylmethyl)-1H-1,2,3-triazolo[4,5-b]pyrazin-6-yl]-1H-pyrazole-1-ethanol mesylate
Chemical Name or Material PF 04217903 mesylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
423910 R & D Systems
RDS423910
10mg Each for $313.93
423950 R & D Systems
RDS423950
50mg Each for $1,298.51
Specifications Description & Specifications

Chemical Identifiers

>98%
468.49
PF 04217903 mesylate
C19H16N8O·CH3SO3H
4-[1-(6-Quinolinylmethyl)-1H-1,2,3-triazolo[4,5-b]pyrazin-6-yl]-1H-pyrazole-1-ethanol mesylate

Specifications

956906-93-7
C19H16N8O·CH3SO3H
4-[1-(6-Quinolinylmethyl)-1H-1,2,3-triazolo[4,5-b]pyrazin-6-yl]-1H-pyrazole-1-ethanol mesylate
PF 04217903 mesylate
>98%
468.49
10mg

Safety and Handling

Recommended Storage : Store at +4°C