Please login to your online account to display your discounted pricing

PF 03814735, Tocris™

Aurora kinase A and B inhibitor

$310.33 - $1,282.40

Chemical Identifiers

Chemical Name or Material PF 03814735
Assay Percent Range >98%
Molecular Formula C23H25F3N6O2
Formula Weight 474.48
Synonym N-[2-[(1S,4R)-6-[[4-(Cyclobutylamino)-5-(trifluoromethyl)-2-pyrimidinyl]amino]-1,2,3,4-tetrahydronaphthalen-1,4-imin-9-yl]-2-oxoethyl]-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

482110

 
r & d systems
482110
10mg Each for $310.33

482150

 
r & d systems
482150
50mg Each for $1,282.40
Description & Specifications

Specifications

Formula Weight 474.48
Quantity 10mg
Chemical Name or Material PF 03814735
Assay Percent Range >98%
Molecular Formula C23H25F3N6O2
CAS 942487-16-3
Synonym N-[2-[(1S,4R)-6-[[4-(Cyclobutylamino)-5-(trifluoromethyl)-2-pyrimidinyl]amino]-1,2,3,4-tetrahydronaphthalen-1,4-imin-9-yl]-2-oxoethyl]-acetamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C