Please login to your online account to display your discounted pricing

Petroleum ether, for residue analysis, 40° to 60°C,ECD tested lindane/DDT<5ng/L, res <0.0003%, ACROS Organics™


Quantity 1L

Manufacturer: thermo fisher scientific  326730010

 View more versions of this product

Catalog No. AC326730010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Petroleum ether
CAS 64742-49-0
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acetic Acid 0.003% max.
RTECS Number OI6180000
EINECS Number 232-453-7
Additional Information GC/ECD (as lindane): max 5ng/L
TSCA TSCA
Purity Grade For residue analysis
Physical Form Liquid
MDL Number MFCD00081849
Merck Index 14, 5489
Synonym Ligroine
Density 0.6500g/mL
Residue after Evaporation 0.0003% max.
Water 0.005% max. (Coulometric)
Boiling Point 40° to 60°C
Color Clear
Quantity 1L

Specifications

Chemical Name or Material Petroleum ether
CAS 64742-49-0
Purity Grade For residue analysis
MDL Number MFCD00081849
Synonym Ligroine
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

SDS & Safety Documents