Learn More
Petroleum Ether, Reagent ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 422995000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Petroleum Ether | |
Passes Test | |
35°C to 60°C | |
â30°C (-86°F) | |
Liquid | |
C6H14 | |
1268 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C | |
86.178 | |
ACS Reagent |
64742-49-0 | |
0.6480g/mL | |
500 mL | |
EcoSafPak™ Glass Bottle | |
0.001% max. | |
MFCD00081849 | |
14, 5489 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
Petroleum ether | |
7892 |
Chemical Identifiers
64742-49-0 | |
86.178 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 | |
CCCC(C)C |
C6H14 | |
MFCD00081849 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
Petroleum ether |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 232-453-7
RTECSNumber : OI6180000
TSCA : TSCA
RUO – Research Use Only