Please login to your online account to display your discounted pricing

Petroleum Ether, Reagent ACS, ACROS Organics™


Quantity 4L
Packaging EcoSafPak™

Manufacturer: thermo fisher scientific  422990040

 View more versions of this product

Catalog No. AC422990040

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Boiling Point 35° to 60°C
Physical Form Liquid
Quantity 4L
Color Clear
Packaging EcoSafPak™
EINECS Number 232-453-7
TSCA TSCA
Density 0.6480g/mL
RTECS Number OI6180000
Merck Index 14, 5489
Chemical Name or Material Petroleum ether
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Residue after Evaporation 0.001% max.
MDL Number MFCD00081849
CAS 64742-49-0
Synonym Benzine
Health Hazard 1 Danger
Acidity Passes Test

Specifications

Chemical Name or Material Petroleum ether
CAS 64742-49-0
Purity Grade ACS Reagent Grade
MDL Number MFCD00081849
Synonym Benzine
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

SDS & Safety Documents