Please login to your online account to display your discounted pricing

Petroleum ether, extra pure, boiling range 40-60°C, ACROS Organics™


Quantity 5L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC124860050

 View more versions of this product

Catalog No. AC124860050

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Boiling Point 40° to 60°C
Physical Form Liquid
Quantity 5L
Color Clear
Merck Index 14, 5489
Packaging Plastic Jerrycan
Density 0.6500g/mL
Chemical Name or Material Petroleum ether
Name Note Extra Dry
Water 0.03% max. (K.F.)
CAS 64742-49-0
Health Hazard 1 Danger
MDL Number MFCD00081849
UN Number 1268
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Residue after Evaporation 0.002% max.
Health Hazard 2 GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

Specifications

Chemical Name or Material Petroleum ether
Name Note Extra Dry
CAS 64742-49-0
Purity Grade Extra Pure
MDL Number MFCD00081849
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

SDS & Safety Documents