Learn More
Petroleum ether, extra pure, boiling range 40-60°C, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 124860025
Specifications
Petroleum ether | |
64742-49-0 | |
MFCD00081849 | |
14, 5489 | |
Solubility in water: insoluble. | |
CCCC(C)C | |
86.178 | |
Liquid | |
0.6500g/mL | |
0.002% max. | |
0.65 | |
−45°C | |
40.0°C to 60.0°C | |
2.5L |
Extra Dry | |
C6H14 | |
1268 | |
1,1-dimethylbutane, 2-methyl pentane, 2-methyl-pentane, 2-methylpentan, dimethylpropylmethane, iso-hexane, isohexane, methyl pentane, pentane, 2-methyl, unii-49ib0u6mld | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
2-methylpentane | |
7892 | |
Extra Pure | |
Glass Bottle | |
Solubility in water: insoluble. | |
0.03% max. (K.F.) | |
0.3 mm2/s (20°C) | |
Undesignated |
Chemical Identifiers
64742-49-0 | |
86.178 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 | |
CCCC(C)C |
C6H14 | |
MFCD00081849 | |
1,1-dimethylbutane, 2-methyl pentane, 2-methyl-pentane, 2-methylpentan, dimethylpropylmethane, iso-hexane, isohexane, methyl pentane, pentane, 2-methyl, unii-49ib0u6mld | |
2-methylpentane |
Safety and Handling
GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger