missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Petroleum ether, extra pure, boiling range 100-140°C, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC233020250
Specifications
Petroleum ether | |
0.5% max. | |
14, 5489 | |
Liquid | |
0.7400g/mL | |
Metal drum | |
0.005% max. | |
0.74 | |
0.76 mm2/s (25°C) | |
Undesignated | |
25L |
64742-49-0,111-65-9,142-82-5,108-87-2,110-82-7,73513-42-5 | |
MFCD00081849 | |
50ppm max. | |
Extra Pure | |
Authentic | |
1.4030 to 1.4080 | |
Solubility in water: insoluble. | |
2°C | |
100.0°C to 140.0°C | |
-30.0°C |