Please login to your online account to display your discounted pricing

Petroleum Ether AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Petroleum Ether
CAS 8032-32-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK49764

 
avantor performance
497608
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 8032-32-4
Packaging Glass
Quantity 4L
Chemical Name or Material Petroleum Ether