missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 180210010

 View more versions of this product

Catalog No. AC180210010




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Petroleum ether
0.6500g/mL
1 L
Glass Bottle
0.65
MFCD00081849
14, 5489
Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil
CCCC(C)C
86.178
ACS Reagent
64742-49-0
40.0°C to 65.0°C
−45°C
1.3660 to 1.3710
C6H14
0.45 mm2/s (25°C)
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
AFABGHUZZDYHJO-UHFFFAOYSA-N
Petroleum ether
7892

Chemical Identifiers

64742-49-0
86.178
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
C6H14
MFCD00081849
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
CCCC(C)C
Safety and Handling

Safety and Handling

GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only