Please login to your online account to display your discounted pricing

Petroleum Ether, ACS, EMD Millipore

$213.17 - $4,509.91

Chemical Identifiers

Chemical Name or Material Petroleum Ether
Name Note For ACS analysis
CAS 8032-32-4
Physical Form Liquid
Synonym Ligroine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX04255

 
emd millipore corporation
PX04255
4L Each for $213.17

MPX04252

 
emd millipore corporation
PX04252
1L Each for $249.97

MPX042520

 
emd millipore corporation
PX042520
200L Each for $4,509.91
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Petroleum Ether
Name Note For ACS analysis
CAS 8032-32-4
Synonym Ligroine