Please login to your online account to display your discounted pricing

Petroleum Ether ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Petroleum Ether
CAS 8032-32-4
Purity Grade ACS Reagent Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4971950

 
avantor performance
497104
AR-CAN 950mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK49804

 
avantor performance
498008
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK498020

 
avantor performance
498019
Steel Pail 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK4980500

 
avantor performance
498004
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK49834

 
avantor performance
498310
SAFEMOR 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Chemical Name or Material Petroleum Ether
Packaging AR-CAN
Quantity 950mL
CAS 8032-32-4