Please login to your online account to display your discounted pricing

Petroleum Ether ACS AR, Macron Fine Chemicals™


macron

Quantity 950mL
Packaging AR-CAN

Manufacturer: avantor performance  497104

 View more versions of this product

Catalog No. MK4971950

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
CAS 8032-32-4
Packaging AR-CAN
Chemical Name or Material Petroleum Ether
Quantity 950mL

Specifications

Chemical Name or Material Petroleum Ether
CAS 8032-32-4
Purity Grade ACS Reagent Grade