Please login to your online account to display your discounted pricing

Petroleum Ether ACS AR, Macron Fine Chemicals™


macron

Quantity 20L
Packaging Steel Pail

Manufacturer: avantor performance  498019

 View more versions of this product

Catalog No. MK498020

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Chemical Name or Material Petroleum Ether
Packaging Steel Pail
Quantity 20L
CAS 8032-32-4

Specifications

Chemical Name or Material Petroleum Ether
CAS 8032-32-4
Purity Grade ACS Reagent Grade