missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Petroleum ether 60/80, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L13734AU

 View more versions of this product

Catalog No. AAL13734AU


Description

Description

It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

Solubility
Insoluble in water.

Notes
Flammable, may be moisture sensitive.
Specifications

Specifications

Petroleum ether 60/80
0.672
1000 mL
Petroleum-like
UN1268
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
8032-32-4
60°C to 80°C
−10°C (14°F)
MFCD00081849
14,5489
Insoluble in water.

Chemical Identifiers

8032-32-4
MFCD00081849
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H340-H350-H304
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.

EINECSNumber : 232-453-7

RTECSNumber : OI6180000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only