Please login to your online account to display your discounted pricing

Petroleum benzine boiling range 40° to 60°C, EMD Millipore

For gas chromatography ECD and FID SupraSolv™

$393.40 - $393.40

Chemical Identifiers

Chemical Name or Material Petroleum benzine
CAS 64742-49-0
Physical Form Liquid
Color Colorless
UN Number UN 1268
Melting Point <−80°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1017722500

 
emd millipore corporation
1017722500
Each for $393.40
Description & Specifications

Specifications

Melting Point <−80°C
Boiling Point 36° to 83°C
Quantity 2.5L
Color Colorless
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
UN Number UN 1268
Odor Benzine-like
Chemical Name or Material Petroleum benzine
Solubility Information Water: 0.01g/L at 20°C soluble
Vapor Pressure 350 hPa (20°C)
CAS 64742-49-0
Health Hazard 1 H225: Highly flammable liquid and vapor.
Ignition Point 250°C (482°F)
DOT Information DOT Class: 3, Packaging Group: II
Density 0.645 to 0.665g/cm3 (15°C)
Physical Form Liquid
EINECS Number 265-151-9
Additional Information SupraSolv™ ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Health Hazard 3 H336: May cause drowsiness or dizziness.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.