Petrolatum (White/Laboratory), Fisher Chemical
Supplier:  Thermo Fisher Scientific P667LB
Catalog No. P66-7LB
Description
Description
Specifications
Specifications
Petrolatum
8009-03-8
White
300°C
C15H15N
7 lb.
WJZSZXCWMATYFX-UHFFFAOYSA-N
1,1,2-trimethyl-1H-benzo[e]indole
approximately 7 mm²/s at 100°C
Paste
White
50°C
0.840g/cm³
Plastic Jar
MFCD00147839
Petroleum Jelly
CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
209.29
Laboratory
Safety and Handling
Safety and Handling
SDS Documents