Please login to your online account to display your discounted pricing

Petrolatum (White/Laboratory), Fisher Chemical


Quantity 1 lb.
Packaging Plastic Jar

Manufacturer: fisher scientific company  P661LB

 View more versions of this product

Catalog No. P66-1LB

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Laboratory Grade
Boiling Point 300°C
Melting Point 50°C
Quantity 1 lb.
Physical Form Paste
Viscosity approximately 7 mm²/s at 100°C
Density 0.840g/cm³
Color White
Identification Pass Test
Packaging Plastic Jar
MDL Number MFCD00147839
Synonym Petroleum Jelly
Chemical Name or Material Petrolatum
Name Note White
CAS 8009-03-8
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Petrolatum
Name Note White
CAS 8009-03-8
Purity Grade Laboratory Grade
Physical Form Paste
MDL Number MFCD00147839
Synonym Petroleum Jelly
Accessories
 

SDS & Safety Documents