Please login to your online account to display your discounted pricing

Petrolatum (White/Laboratory), Fisher Chemical

$311.38 - $1,208.00

Chemical Identifiers

Chemical Name or Material Petrolatum
Name Note White
CAS 8009-03-8
Purity Grade Laboratory Grade
Physical Form Paste
MDL Number MFCD00147839
Synonym Petroleum Jelly
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P66-7LB

  SDS
fisher scientific company
P667LB
Plastic Jar 7 lb. Each for $1,208.00
Case of 4 Each for $4,832.00

P66-1LB

  SDS
fisher scientific company
P661LB
Plastic Jar 1 lb. Each for $311.38
Case of 6 Each for $1,868.28
Description & Specifications

Specifications

Purity Grade Laboratory Grade
Boiling Point 300°C
Melting Point 50°C
Quantity 7 lb.
Physical Form Paste
Density 0.840g/cm³
Viscosity approximately 7 mm²/s at 100°C
Color White
Synonym Petroleum Jelly
Identification Pass Test
Packaging Plastic Jar
MDL Number MFCD00147839
CAS 8009-03-8
ChemAlert Storage Symbol Gray
Chemical Name or Material Petrolatum
Name Note White

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling