Please login to your online account to display your discounted pricing

Peterson First Guides™


Guides for the beginning naturalist

Manufacturer: fisher science education  0395911834

 View more versions of this product

Catalog No. S32092

  • / Each
 

Description & Specifications

Specifications

Product Type Peterson First Guides: Trees
Author Janet Wehr, Olivia Petrides, George Petrides, Roger Peterson
Category Biology
For Use With (Application) Focuses on the animals, plants and other natural things, Each contains full-color illustrations and clear descriptions
Format Paperback