Please login to your online account to display your discounted pricing

Peterson First Guides™


Guides for the beginning naturalist

Manufacturer: fisher science education  0395906660

 View more versions of this product

Catalog No. S32083

  • / Each
 

Description & Specifications

Specifications

Author Roger Peterson, Roger Peterson
Category Biology
For Use With (Application) Focuses on the animals, plants and other natural things, Each contains full-color illustrations and clear descriptions
Format Paperback
Product Type Peterson First Guides: Birds of North America