missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pesticides Surrogate Standard Spiking Solution, 200μg/mL in acetone, Ultra Scientific

$86.49 - $86.49

Chemical Identifiers

CAS 000067-64-1,000877-09-8,002051-24-3
CAS Max % 99.95,0.02,0.02
Chemical Name or Material Pesticides Surrogate Standard Spiking Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
US-ISM-3201 Ultra Scientific
801175018933
1 x 1mL Each for $86.49
Specifications Description & Specifications

Chemical Identifiers

000067-64-1,000877-09-8,002051-24-3
99.95,0.02,0.02
Pesticides Surrogate Standard Spiking Solution

Specifications

1 x 1mL
Acetone,2,4
99.95,0.02,0.02
Pesticides Surrogate Standard Spiking Solution
Acetone
000067-64-1,000877-09-8,002051-24-3
200μg/mL