Pesticides Resolution Check Mixture, Various Concentrations in Hexane, Ultra Scientific
Manufacturer:  Ultra Scientific CLP2421
 View more versions of this product
Catalog No. USCLP2421
Specifications Description & Specifications
Specifications
Pesticides Resolution Check Mixture
Liquid
1 x 1mL
000110-54-3,005103-74-2,000959-98-8,000072-55-9,000060-57-1,001031-07-8
gamma-chlordane: 10ng/mL; endosulfan I: 10ng/mL; 4,4apostrophe-DDE: 20ng/mL; dieldrin: 20ng/mL; endosulfan sulfate: 20ng/mL; endrin ketone: 20ng/mL; methoxychlor: 100ng/mL; 2,4,5,6-tetrachloro-m-xylene: 20ng/mL; decachlorobiphenyl (BZ No. 209):
Chemical Identifiers
000110-54-3,005103-74-2,000959-98-8,000072-55-9,000060-57-1,001031-07-8
Liquid
Pesticides Resolution Check Mixture