missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pesticides Resolution Check Mixture, Various Concentrations in Hexane, Ultra Scientific

$107.66 - $107.66

Chemical Identifiers

CAS 000110-54-3,005103-74-2,000959-98-8,000072-55-9,000060-57-1,001031-07-8
Physical Form Liquid
Chemical Name or Material Pesticides Resolution Check Mixture
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
USCLP2421 Ultra Scientific
CLP2421
1 x 1mL Each for $107.66
Specifications Description & Specifications

Chemical Identifiers

000110-54-3,005103-74-2,000959-98-8,000072-55-9,000060-57-1,001031-07-8
Liquid
Pesticides Resolution Check Mixture

Specifications

000110-54-3,005103-74-2,000959-98-8,000072-55-9,000060-57-1,001031-07-8
gamma-chlordane: 10ng/mL; endosulfan I: 10ng/mL; 4,4apostrophe-DDE: 20ng/mL; dieldrin: 20ng/mL; endosulfan sulfate: 20ng/mL; endrin ketone: 20ng/mL; methoxychlor: 100ng/mL; 2,4,5,6-tetrachloro-m-xylene: 20ng/mL; decachlorobiphenyl (BZ No. 209):
Pesticides Resolution Check Mixture
Liquid
1 x 1mL