missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Rimsulfuron, MilliporeSigma™ Supelco™

Catalog No. 111012811
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012811 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012811 Supplier Merck Emd Millipore Supplier No. 31658100MG
Only null left

Rimsulfuron is a sulfonylurea herbicide primarily used to control a wide variety of perennial and annual grasses and some broadleaf weeds. The mode of action of rimsulfuron involves inhibition of acetolactate synthase (ALS), a key enzyme in the biosynthetic pathway of branched-chain amino acids.

Agriculture

Specifications

Product Line PESTANAL
CAS 122931-48-0
Flash Point >200°C
Physical Form Neat
Linear Formula C14H17N5O7S2
MDL Number MFCD01632305
Synonym 1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(ethylsulfonyl)-2-pyridylsulfonyl]urea
Type Analytical Standard
Quantity 100 mg
Formula Weight 431.44
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH