missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cyanuric acid-13C3, MilliporeSigma™ Supelco™

Catalog No. 111013453
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013453 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013453 Supplier Merck Emd Millipore Supplier No. 3267910MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 201996-37-4
Physical Form Neat
Linear Formula 13C3H3N3O3
MDL Number MFCD00209704
Melting Point >360°C
Synonym 1,3,5-Triazine-2,4,6-triol-13C3; 2,4,6-Trihydroxy-1,3,5-triazine-13C3
Type Analytical Standard
Quantity 10 mg
Formula Weight 132.05
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH