missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Benfuresate, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 68505-69-1 |
| Physical Form | Neat |
| Linear Formula | C12H16O4 S |
| MDL Number | MFCD01728672 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 256.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH