missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Atrazine-desisopropyl-2-hydroxy, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 7313-54-4 |
| Physical Form | Neat |
| Linear Formula | C5H9N5 O |
| MDL Number | MFCD01310649 |
| Synonym | 6-Amino-4-ethylamino-2-hydroxy-1,3,5-triazine |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 155.16 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH