missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 2-Imidazolidinethione-4,4,5,5-d4, ≥97.5% (GC), MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 352431-28-8 |
| Linear Formula | C3D4H2N2 S |
| MDL Number | MFCD01317470 |
| Synonym | 2-Thioxoimidazolidine-4,4,5,5-d4; N,N′-Ethylene-d4-thiourea |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 106.18 |
| Percent Purity | ≥97.5% (GC) |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH