missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perylene, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130070010

 View more versions of this product

Catalog No. AC130070010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Perylene
277.0°C to 280.0°C
Authentic
Glass bottle
MFCD00004142
05, I, 363
peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl
CSHWQDPOILHKBI-UHFFFAOYSA-N
perylene
9142
252.31
Powder
198-55-0
Ochre to Brown
99% min. (HPLC)
C20H12
1 g
15, 7299
Solubility in water: insoluble. Other solubilities: slightly soluble in ether, alcohol,, soluble in benzene, toluene,, very soluble in cs2, chloroform, acetone, very sparingly soluble in petroleum ether
C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2
252.31
CHEBI:29861
99+%

Chemical Identifiers

198-55-0
252.31
CSHWQDPOILHKBI-UHFFFAOYSA-N
9142
perylene
C20H12
MFCD00004142
peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl
CHEBI:29861
C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2
Safety and Handling

Safety and Handling

EINECSNumber : 205-900-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only