Please login to your online account to display your discounted pricing

Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  316180050

 View more versions of this product

Catalog No. AC316180050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 251.21
Purity Grade Trace Metal Basis
Quantity 5g
Color Clear colorless to pale yellow
Density 2.1600g/mL
Name Note 99.99%
Concentration 70 to 76.5% (HReO4)
Linear Formula HReO4
MDL Number MFCD00011326
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material Perrhenic(VII)acid
Total Trace Metal Impurities 100ppm max.
Assay Percent Range 99.99%
Molecular Formula HO4Re
CAS 13768-11-1
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material Perrhenic(VII)acid
Name Note 99.99%
CAS 13768-11-1
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula HO4Re
Linear Formula HReO4
Formula Weight 251.21
MDL Number MFCD00011326
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents