missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perrhenic acid, 75-80% aq. soln., Alfa Aesar™

Manufacturer:  Alfa Aesar 1141306

 View more versions of this product

Catalog No. AA1141306



Specifications

Specifications

Colorless to pale yellow liquid
23 hPa (17mm Hg) at 20°C
5g
75-80%
MFCD00011326
Perrhenic acid: 80.0%; Water: 20.0%
O[Re](=O)(=O)=O
251.211
251.26
Perrhenic acid
Colorless to Yellow
13768-11-1
HO4Re
acmc-1bqjs
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
hydroxy(trioxo)rhenium
83718
Liquid

Chemical Identifiers

13768-11-1
251.211
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
83718
O[Re](=O)(=O)=O
HO4Re
MFCD00011326
acmc-1bqjs
hydroxy(trioxo)rhenium
Safety and Handling

Safety and Handling

H314-H318-H290-H302: Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 237-380-4

Documents