missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Peroxidase from Horseradish, TCI America™

Manufacturer:  TCI America P00731G

 View more versions of this product

Catalog No. P00731G



Specifications

Specifications

Peroxidase from Horseradish
1g
MFCD00071339
Undesignated
9003-99-0
Crystal-Powder at 20°C

Chemical Identifiers

9003-99-0
MFCD00071339
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer

Documents