missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Peroxidase from Horseradish, TCI America™

$224.59 - $1,223.78

Chemical Identifiers

CAS 9003-99-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P0073100MG View Documents TCI America
P0073100MG
100mg Each for $224.59
 
P00731G View Documents TCI America
P00731G
1g Each for $1,223.78
 
Specifications

Chemical Identifiers

9003-99-0

Specifications

Undesignated
100mg
9003-99-0
Crystal - Powder
Safety and Handling

Safety and Handling

Recommended Storage : <0°C