Please login to your online account to display your discounted pricing

Periodic Acid AR, Macron Fine Chemicals™


macron

Quantity 25g
Packaging Glass

Manufacturer: avantor performance  E42255

 View more versions of this product

Catalog No. MKE42225

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 227.94
Chemical Name or Material Periodic Acid
Name Note Dihydrate
Packaging Glass
Molecular Formula HIO4·2H2O
CAS 10450-60-9
Quantity 25g

Specifications

Chemical Name or Material Periodic Acid
Name Note Dihydrate
CAS 10450-60-9
Molecular Formula HIO4·2H2O
Formula Weight 227.94