missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Periodic acid, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC453170250
Specifications
Periodic Acid | |
10450-60-9 | |
H5IO6 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
pentahydroxy(oxo)-$l^{7}-iodane | |
25289 | |
227.94 | |
Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol, slightly soluble in ether |
25g | |
H5IO6 | |
acid, periodic, hexaoxoiodic 5-acid, orthoperiodic acid, paraperiodic acid, pentahydroxidooxidoiodine, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, tetrahydroxyiodous acid, unii-ak1d44l87g | |
OI(=O)(O)(O)(O)O | |
227.938 | |
CHEBI:29150 | |
Analysis | |
122.0°C |
Chemical Identifiers
10450-60-9 | |
227.938 | |
acid, periodic, hexaoxoiodic 5-acid, orthoperiodic acid, paraperiodic acid, pentahydroxidooxidoiodine, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, tetrahydroxyiodous acid, unii-ak1d44l87g | |
CHEBI:29150 | |
OI(=O)(O)(O)(O)O |
H5IO6 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
25289 | |
pentahydroxy(oxo)-$l^{7}-iodane |