Learn More
Periodic acid, 99%
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Supplier: Thermo Scientific Chemicals B2043318
Description
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPeriodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
Solubility
Soluble in water and alcohols.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, strong bases and dimethyl sulfoxide.
Specifications
| 1.2 | |
| 126°C to 130°C | |
| Odorless | |
| 50 g | |
| MFCD00011344 | |
| 14,7171 | |
| Soluble in water and alcohols. | |
| O[I](O)(O)(O)(O)=O | |
| 227.94 | |
| CHEBI:29150 | |
| 99% |
| Periodic acid | |
| 10450-60-9 | |
| Hygroscopic; Light sensitive | |
| H5IO6 | |
| UN3085 | |
| orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| pentahydroxy(oxo)-$l^{7}-iodane | |
| 25289 | |
| 227.94 |
Chemical Identifiers
| 10450-60-9 | |
| 227.94 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| 25289 | |
| pentahydroxy(oxo)-$l^{7}-iodane |
| H5IO6 | |
| MFCD00011344 | |
| orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
| CHEBI:29150 | |
| O[I](O)(O)(O)(O)=O |
Safety and Handling
GHS H Statement
H272-H314
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
H271-H314-H335-H372
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber : 233-937-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only