missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Periodic acid, 99+%, ACROS Organics™

Manufacturer:  Acros Organics 198400050

 View more versions of this product

Catalog No. AC198400050



Specifications

Specifications

Periodic acid
1.0% max. Iodic Acid
H5IO6
01,815; 02,313; 03,220; 04,374; 05,508; 10,304; 13,238; 16,272
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
OI(=O)(O)(O)(O)O
227.938
CHEBI:29150
Crystals or Crystalline Powder
200ppm max. (Bromide, Bromate (as Cl))
Positive for periodate
10ppm max.
100ppm max.
0.05% max.
5g
10450-60-9
99+%
H5IO6
15, 7286
TWLXDPFBEPBAQB-UHFFFAOYSA-N
pentahydroxy(oxo)-$l^{7}-iodane
25289
227.94
≥99.0% (Iodometry)
200ppm max. (as Cl)
0.01% max.
50ppm max.
200ppm max.
White to Yellow

Chemical Identifiers

10450-60-9
227.938
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
CHEBI:29150
OI(=O)(O)(O)(O)O
H5IO6
TWLXDPFBEPBAQB-UHFFFAOYSA-N
25289
pentahydroxy(oxo)-$l^{7}-iodane
Safety and Handling

Safety and Handling

Danger

SDS Documents