missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pergolide, TCI America™

Manufacturer:  TCI America P2200200MG

 View more versions of this product

Catalog No. P2200200MG



Specifications

Specifications

Pergolide
210°C
66104-22-1
MFCD00867357
YEHCICAEULNIGD-MZMPZRCHSA-N
(6aR,9R,10aR)-9-(methylsulfanylmethyl)-7-propyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline
47811
314.49
≥97.0% (HPLC)
White-Yellow
200mg
C19H26N2S
1544
CCCN1CC(CC2C1CC3=CNC4=CC=CC2=C34)CSC
314.491
CHEBI:63617
Crystal-Powder at 20°C

Chemical Identifiers

66104-22-1
314.491
YEHCICAEULNIGD-MZMPZRCHSA-N
CHEBI:63617
CCCN1CC(CC2C1CC3=CNC4=CC=CC2=C34)CSC
C19H26N2S
MFCD00867357
47811
(6aR,9R,10aR)-9-(methylsulfanylmethyl)-7-propyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline
Safety and Handling

Safety and Handling

RTECSNumber : KE6344964

TSCA : No

Recommended Storage : Freezer (-20 °C)

Documents